

Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®
JT9012-3
:
- Pack type:Glass bottle, amber, narrow mouth
- Size:4 L
- Cat. No.:JT9012-3
- Supplier no.:9012-03
- Appearance:Passes Test
- Assay (GC):≥99.9 %
- Color (APHA):≤10
- Water (H₂O):≤0.02 %
- Gradient Elution Test (a.u.) - 254 nm:≤0.005
- Residue after Evaporation:≤3 ppm
- Titrable Acid (µeq/g):≤8
- Titrable Base (µeq/g):≤0.6
- Ultraviolet Absorbance (1.00 cm) - 200 nm:≤0.1
- Ultraviolet Absorbance (1.00 cm) - 210 nm:≤0.05
- Ultraviolet Absorbance (1.00 cm) - 220 nm:≤0.03
- Ultraviolet Absorbance (1.00 cm) - 254 nm:≤0.01
- Ultraviolet Absorbance (1.00 cm) - 280 nm:≤0.01
- Ultraviolet Absorbance (1.00 cm) - 350 nm:≤0.01
- Ultraviolet Absorbance (1.00 cm) - 400 nm:≤0.01
- Ultraviolet Absorbance (1.00 cm) - UV Cut-off, nm:≤190
:
:41.05 g/mol
:13,00071
:81.6 °C (1013 hPa)
:–45.7 °C
:0.782 g/cm³ (20 °C)
:2 °C (closed cup)
:Ambient
:200-835-2
:1648
:3,II
:Management of Change (MOC) category = N
:01-2119471307-38
:GHS02
:GHS07
Frequently Bought Together