Order Entry
United States
Orders LinkContactUsLinkComponent
2-Cyclohexylethanol 99%

2-Cyclohexylethanol 99%

Bottle, 250 g
Catalog # AAB20782-30
Irritant
Supplier #:  B20782-30
2-Cyclohexylethanol 99%
Catalog # AAB20782-30
CAS Number:  4442-79-9
2-Cyclohexylethanol 99%
2-Cyclohexylethanol 99%

Bottle, 250 g

CAS Number:  4442-79-9
Price
$146.60
Log in for availability
Each (250g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 99% CAS No.: 4442-79-9 Molecular Formula: C8H16O Formula Weight: 128.22 Flash Point: 86°(186°F) Refractive Index: 1.4660 Melting Point: -20° Boiling Point: 206-207° Beilstein Reference: 1848152 MDL No.: MFCD00001525

  • CAS Number:4442-79-9
  • Formula:C₈H₁₆O
  • MDL Number:MFCD00001525
  • Molecular Weight:128.21 g/mol
  • Boiling Pt:206…207 °C
  • Melting Pt:–20 °C
  • Density:0.920
  • Flash Pt:86 °C(186 °F)
  • Storage Temperature:Ambient
  • EINECS:224-672-1
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    250 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.