Order Entry
United States
Orders LinkContactUsLinkComponent
Ammonia solution standardised solution 5.0 N

Ammonia solution standardised solution 5.0 N

1 L
Catalog # AA35577-K2
Corrosive
Irritant
Supplier #:  035577-K2
Ammonia solution standardised solution 5.0 N
Catalog # AA35577-K2
CAS Number:  1336-21-6
Ammonia solution standardised solution 5.0 N

1 L

CAS Number:  1336-21-6
Price
$106.33
Log in for availability
Each (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS No.: 1336-21-6 Molecular Formula: H5NO Formula Weight: 35.05 Physical Form: Liquid Appearance: Colorless Odor: Ammonia-like UN No.: 2672 MDL No.: MFCD00066650

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III

Documents

Specifications

Specifications

  • Size:
    1 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.