Orders LinkContactUsLinkComponent
Iron(III) hydroxide, γ-phase
50 g
  AA17531-18
 :  017531-18
Iron(III) hydroxide, γ-phase
  AA17531-18
 :  20344-49-4
Iron(III) hydroxide, γ-phase
50 g
 :  20344-49-4
$3,168.64
Each (50g)
CAS No.: 20344-49-4 Molecular Formula: FeO(OH) Formula Weight: 88.85 Physical Form: -60 Mesh Powder Odor: Odorless Melting Point: 135° dec. MDL No.: MFCD00064782
  •  :20344-49-4
  •  :FeO(OH)
  •  :MFCD00064782
  •  :88.85 g/mol
  •  :14,4025
  •  :135 °C (dec.)
  •  :Ambient
  •  :243-746-4
  • Size:
    50 g