Order Entry
United States
Orders LinkContactUsLinkComponent
Triethylamine 99+%

Triethylamine 99+%

Bottle, 1 kg
Catalog # AA12391-A1
Flammable
Corrosive
Irritant
Supplier #:  012391-A1
Triethylamine 99+%
Catalog # AA12391-A1
CAS Number:  121-44-8
Triethylamine 99+%
Triethylamine 99+%

Bottle, 1 kg

CAS Number:  121-44-8
Price
$111.43
Log in for availability
Each (1kg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL: MFCD00009051 Beilstein Registry No.: 1843166 Fieser: 1,1198 2,427 4,527 5,689 7,385 9,481 15,336

Purity: 99+% CAS No.: 121-44-8 Molecular Formula: C6H15N Formula Weight: 101.19 Flash Point: -11°(12°F) Physical Form: Liquid Density: 0.726 Refractive Index: 1.4005 Appearance: Colorless Odor: Ammonia-like Melting Point: -115° Boiling Point: 89-90° Beilstein Reference: 1843166 Merck Reference: 14,9666 UN No.: 1296 MDL No.: MFCD00009051

Synonyms:  (Diethylamino)ethane, TEA
  • CAS Number:121-44-8
  • Formula:(C₂H₅)₃N
  • MDL Number:MFCD00009051
  • Molecular Weight:101.19 g/mol
  • Merck Index:14,9666
  • Boiling Pt:89…90 °C
  • Melting Pt:–115 °C
  • Density:0.726
  • Flash Pt:–11 °C (–12 °F)
  • Storage Temperature:Ambient
  • EINECS:204-469-4
  • UN:1296
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    1 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.