Orders LinkContactUsLinkComponent
Draft Shields for Equipments
Draft Shields for Equipments
Draft shield with sliding doors, 1 mg, 168 mm
Catalog # 97044-172
Supplier:   Mettler Toledo
Supplier #:  12122405
Draft Shields for Equipments
Catalog # 97044-172
Supplier:  Mettler Toledo
Draft Shields for Equipments
Draft Shields for Equipments
Draft shield with sliding doors, 1 mg, 168 mm
Price
$1,130.00
Log in for availability
Each
Product Details & Documents
The draft shield minimizes air turbulence around your balance, enabling results to be delivered faster.
  • Faster stabilization time
  • Speed up your weighing processes
Specifications
  • Description:
    Draft shield with sliding doors, 1 mg
  • Height:
    168 mm