Order Entry
United States
Orders LinkContactUsLinkComponent
Hardness 1 Buffer Solution, Hach

Hardness 1 Buffer Solution, Hach

Hardness 1 Buffer Solution, 1 L, EDTA, Bottle
Catalog # 97026-632
Product available on GSA Advantage®
Supplier:   Hach
Supplier #:  42453
Hardness 1 Buffer Solution, Hach
Catalog # 97026-632
Supplier:  Hach
Product available on GSA Advantage®
CAS Number:  
Hardness 1 Buffer Solution, Hach
Hardness 1 Buffer Solution, Hach

Hardness 1 Buffer Solution, 1 L, EDTA, Bottle

CAS Number:  
Price
$136.20
Log in for availability
Each
Product Details & Documents

Buffer for hardness and chelant determination by titration, pH 10.1 ± 0.1 at 20°C

Hach methods 8213 and 8226. Also used in Hach Phenols method 8047.

Documents

Specifications

Specifications

  • Chemical method:
    EDTA
  • Pack type:
    Bottle
  • Description:
    Hardness 1 Buffer Solution
  • Size:
    1 L
  • Cat. No.:
    97026-632
  • Method:
    EDTA Titration

Specifications