Corning® PureCoat™ Flasks, Amine, Corning
75 cm2, 5 Plates/Pack, 50 Packs/Case
Supplier: Corning
Supplier #: 356726
Jump to:
Product Details & Documents
PS, with blue screw cap.
Corning® PureCoat™ amine flasks provide an enhanced tissue culture surface. The positively charged surface, manufactured using a proprietary thin-film coating technology, offers improved attachment, increased proliferation, and enhanced recovery from freeze-thaw for a broad range of primary, transfected and fastidious cells that exhibit poor attachment properties on standard tissue culture (TC) surfaces in serum-free/serum-reduced conditions.
Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Documents
Specifications
- Size:75 cm2
- Packed:5 Plates/Pack, 50 Packs/Case