Order Entry
United States
Orders LinkContactUsLinkComponent
Dehydrant Blends, Epredia

Dehydrant Blends, Epredia

Dehydrant 95, Metal Drum, 55 gal.
Catalog # 84000-048
Supplier:   Epredia
Supplier #:  6355
Dehydrant Blends, Epredia
Catalog # 84000-048
Supplier:  Epredia
CAS Number:  
Dehydrant Blends, Epredia
Dehydrant Blends, Epredia

Dehydrant 95, Metal Drum, 55 gal.

CAS Number:  
Price
$1,458.68
New Customer Promotional Price
Log in for availability
Each (55US Gal)
Product Details & Documents

Process and stain histological and cytological specimens with these blends of denatured ethyl alcohol reagent.

  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents

Process and stain histological and cytological specimens with Epredia™ Dehydrant Blends. These denatured ethyl alcohol reagent blends are specifically designed for processing and staining histological and cytological specimens.

Specifications

Specifications

  • Pack type:
    Metal Drum
  • Description:
    Dehydrant 95
  • Size:
    55 gal.
  • Application:
    Processing and Staining Histological and Cytological Specimens

Specifications