Proto® Puller Step Plates, ORS Nasco
Puller Step Plate, 1 – 11/4", 0.11 lbs.
300024-964
: 577-4040-3
Provides forcing screw base for hollow shaft applications 3/4 to 23/8" bore.
- Description:Puller Step Plate
- Weight:0.11 lbs.
- Diameter:1 – 11/4"