Order Entry
United States
Orders LinkContactUsLinkComponent
Tissue-Tek® Processing/Embedding Cassette Systems, Sakura

Tissue-Tek® Processing/Embedding Cassette Systems, Sakura

Orange
Catalog # 25608-790
Supplier:   Sakura Finetek
Supplier #:  4126
Tissue-Tek® Processing/Embedding Cassette Systems, Sakura
Catalog # 25608-790
Supplier:  Sakura Finetek
Tissue-Tek® Processing/Embedding Cassette Systems, Sakura
Tissue-Tek® Processing/Embedding Cassette Systems, Sakura

Orange

Price
$290.00
Log in for availability
Case of 1,500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Cassette systems (cassette, cover, and base) are designed for processing, embedding, sectioning, and storage

Reusable base molds enable perfect specimen positioning. Extended fins prevent tipping and provide gripping surface. Stainless steel construction ensures uniform heating and cooling.

Note: Clear cassettes are polypropylene and are not recommended for use in processors using heat or vacuum.

Plastic cassettes feature an angled, textured writing surface. Available in clear and in 10 colors.

Ordering information: Reusable stainless steel cover sold separately.

Specifications
  • Color:
    Orange
Accessories

Product Information

  • Catalog #
    Length
    Width
    Availability
    Price
  • Base Mold
    Base Mold
    25608-826
    15 mm
    15 mm
    Log in for availability
    Case of 12 - $116.00

Product Information

  • Catalog #
    Length
    Width
    Availability
    Price
  • Base Mold
    Base Mold
    25608-832
    37 mm
    24 mm
    Log in for availability
    Case of 12 - $116.00

Product Information

  • Catalog #
    Length
    Width
    Availability
    Price
  • Base Mold
    Base Mold
    25608-830
    30 mm
    24 mm
    Log in for availability
    Case of 12 - $116.00

Product Information

  • Catalog #
    Length
    Width
    Availability
    Price
  • Base Mold
    Base Mold
    25608-824
    7 mm
    7 mm
    Log in for availability
    Case of 12 - $116.00

Product Information

  • Catalog #
    Availability
    Price
  • Stainless Steel Cover
    Stainless Steel Cover
    25608-870
    Log in for availability
    Case of 25 - $70.50

Product Information

  • Catalog #
    Length
    Width
    Availability
    Price
  • Base Mold
    Base Mold
    25608-828
    24 mm
    24 mm
    Log in for availability
    Case of 12 - $116.00

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.