Orders LinkContactUsLinkComponent
Nunc® CryoColor Coder Inserts, Thermo Scientific
Nunc® CryoColor Coder Inserts, Thermo Scientific
White
Catalog # 24270-100
Supplier #:  354755
Nunc® CryoColor Coder Inserts, Thermo Scientific
Catalog # 24270-100
Nunc® CryoColor Coder Inserts, Thermo Scientific
Nunc® CryoColor Coder Inserts, Thermo Scientific
White
Price
$85.24
New Customer Promotional Price
Log in for availability
Pack of 500 Case of 2000
Product Details & Documents
For comprehensive sample identification, fits inside CryoTube® vial cap.
  • Made of polystyrene
  • Available in 10 different colors
Ordering information: Assortment 24270-120 includes either 50 of each color (for a pack) or 200 of each color (for a case).
Specifications
  • Color:
    White