Order Entry
United States
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB), liquid substrate

3,3',5,5'-Tetramethylbenzidine (TMB), liquid substrate

100 ml
Catalog # IC15234680
Irritant
Supplier:   MP Biomedicals
Supplier #:  0215234680
3,3',5,5'-Tetramethylbenzidine (TMB), liquid substrate
Catalog # IC15234680
Supplier:  MP Biomedicals
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB), liquid substrate
3,3',5,5'-Tetramethylbenzidine (TMB), liquid substrate

100 ml

CAS Number:  54827-17-7
Price
$187.00
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

TMB is a chromogenic substrate for Horse Radish Peroxidase (HRP). MP Biolomedicals TMB Solution is a convenient, non toxic and ready to use stable chromogen solution suitable for peroxidase and pseudo-peroxidase reactions in ELISA, it provides consistent performance and sensitivity to deliver reproducible results

  • Ready-to-Use, convenient
  • Reproducible Results
  • Consistent Performance and Sensitivity
  • Stabilized chromogen solution
  • For ELISA procedures and peroxidase and pseudo-peroxidase reactions
  • Stable up to 24 months
  • Non toxic.

Metabolism of the TMB substrate by HRP creates a soluble bluish reaction product that may be quantified by measuring absorbance at 370 or 665 nm. Reaction can be stopped by addition of 1 volume of 1N hydrocloric or sulfurc acid to yield a yellow reaction product that can be measured by absorbance at 450 nm (within one hour).

Caution: Research Use Only (RUO).

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Functionality Test :
    Passes
  • pH:
    3.1 ± 0.5
  • Expiration Date:
    Month/Day/Year

Specifications

  • Size:
    100 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.