Order Entry
United States
Orders LinkContactUsLinkComponent
Valinomycin ≥93%, solid

Valinomycin ≥93%, solid

10 mg
Catalog # IC10501010
Supplier:   MP Biomedicals
Supplier #:  0210501010
Valinomycin ≥93%, solid
Catalog # IC10501010
Supplier:  MP Biomedicals
CAS Number:  2001-95-8
Valinomycin ≥93%, solid
Valinomycin ≥93%, solid

10 mg

CAS Number:  2001-95-8
Price
$101.00
Log in for availability
Each (10mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

A metabolite with a broad antibiotic spectrum

  • Presentation: White
  • Freely soluble in petroleum ether, ether, benzene, chloroform, glacial acetic, butyl acetate; soluble in methanol (5 mg/ml-clear, colorless solution) and DMSO (≥10 mg/mL); practically insolubable in water.

Valinomycin, is used as a potassium ionophore shown to induce cell death. Due to the ion-selective nature of valinomycin, it is used in ion-selective electrodes.

K+-selective ionophoric cyclodepsipeptide; potassium ionophore which uncouples oxidative phosphorylation, induces apoptosis in murine thymocytes, inhibits NGF-induced neuronal differentiation and antagonizes ET-induced vasoconstriction.

Inhibitor of mitochondrial action. Valinomycin is a cyclododecadepsipeptide potassium-selective ionophore. It is an antibiotic that was originially isolated from various strains of Streptomyces. The hydrophilic interior is the right size to accommodate the potassium ion, but not other ions, while the hydrophobic exterior allows the complex to pass through the lipid bilayer. Valinomycin induces apoptosis in several cell types, including CHO cells, by stimulating potassium efflux. Apoptotic events produced by valinomycin include phosphatidylserine membrane translocation, caspase-3 activation, and mitochondrial membrane depolarization. Due to the ion-selective nature of valinomycin, it is used in ion-selective electrodes.

White crystalline powder

Freely soluble in petroleum ether, ether, benzene, chloroform, glacial acetic, butyl acetate; soluble in methanol (5 mg/mL-clear, colorless solution) and DMSO (≥10 mg/mL); practically insolubable in water

  • CAS Number:2001-95-8
  • Formula:C₅₄H₉₀O₁₈N₆
  • MDL Number:MFCD00005114
  • Molecular Weight:1111.34 g/mol
  • Melting Pt:184…190 °C
  • Storage Temperature:Refrigerator
  • ADR:6.1,I
Specifications

Specifications

  • Size:
    10 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.