Order Entry
United States
Orders LinkContactUsLinkComponent
Universal® Multi Purpose Paper

Universal® Multi Purpose Paper

Unpunched papers, Bright white, 8.5×14", 20 lbs.
Catalog # 500020-269
Marketsource
Supplier #:  UNV95400
Universal® Multi Purpose Paper
Catalog # 500020-269
Supplier:  
Marketsource
Universal® Multi Purpose Paper
Universal® Multi Purpose Paper

Unpunched papers, Bright white, 8.5×14", 20 lbs.

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PAPER XERO/DUP LTR 98B WE CT10 Catalog #500020-270

Product 500020-270 is similar to 500020-269 but manufacturer and specifications may vary.

Product Details & Documents

Perfect for all Laser Printers, Copiers, Fax Machines and Multifunction Machines.

  • Brightness rating 108 (International) and 98 (US)
  • Acid-free for archival quality

Now with Colorlok™ for enhanced colors and sharper text and make your documents stand out.

Specifications
  • Color:
    Bright white
  • Description:
    Unpunched papers
  • Dimensions:
    8.5×14"
  • Weight:
    20 lbs.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.