Order Entry
United States
Orders LinkContactUsLinkComponent
Pacon® Array® Card Stock

Pacon® Array® Card Stock

Parchment color card stock, assorted colors (blue, gold, natural, pink, tan), Assorted, 8.5×11", 65 lbs.
Catalog # 500004-261
Marketsource
Supplier:   Janitorial Supplies
Supplier #:  PAC101235
Pacon® Array® Card Stock
Catalog # 500004-261
Marketsource
Pacon® Array® Card Stock
Pacon® Array® Card Stock

Parchment color card stock, assorted colors (blue, gold, natural, pink, tan), Assorted, 8.5×11", 65 lbs.

Price
Inquire For Price
Log in for availability
Pack of 100
Minimum order quantity is 1
Product Details & Documents

For report covers, flyers, postcards and art projects.

  • Acid-free for archival quality

Used in printers, copiers or typewriters.

Specifications
  • Color:
    Assorted
  • Description:
    Parchment color card stock, assorted colors (blue, gold, natural, pink, tan)
  • Dimensions:
    8.5×11"
  • Weight:
    65 lbs.