Order Entry
United States
Orders LinkContactUsLinkComponent
Avery® Durable Slant Ring Binder

Avery® Durable Slant Ring Binder

Binder without Spine Inserts, 3", 600, Black, Nonlocking Rings, Vinyl, 11 x 8 1/2"
Catalog # 500001-550
Marketsource
Avery® Durable Slant Ring Binder
Catalog # 500001-550
Supplier:  
Marketsource
Avery® Durable Slant Ring Binder
Avery® Durable Slant Ring Binder

Binder without Spine Inserts, 3", 600, Black, Nonlocking Rings, Vinyl, 11 x 8 1/2"

Discontinued
Product Details & Documents

Gap Free™ feature prevents ring misalignment so pages turn smoothly.

  • Four interior pockets (two back and two front) store unbound materials so nothing gets lost
  • Three slant ring fasteners are used
  • Non view binders

Back-mounted slant rings hold up to 25% more sheets than same size round rings and allow pages to lie flat.

Specifications
  • Sheet capacity:
    600
  • Capacity:
    3"
  • Color:
    Black
  • Description:
    Binder without Spine Inserts
  • Material:
    Vinyl
  • Size:
    11 x 8 1/2"
  • Style:
    Nonlocking Rings