Orders LinkContactUsLinkComponent
Sure Shot® Spray-Safe Atomizer, Electron Microscopy Sciences
Sure Shot® Spray-Safe Atomizer, Electron Microscopy Sciences
Compete Repair Kit
Catalog # 101412-904
Supplier #:  64189
Sure Shot® Spray-Safe Atomizer, Electron Microscopy Sciences
Catalog # 101412-904
Sure Shot® Spray-Safe Atomizer, Electron Microscopy Sciences
Sure Shot® Spray-Safe Atomizer, Electron Microscopy Sciences
Compete Repair Kit
Price
$29.77
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sure Shot® pressure sprayers are available in 16, 8 and 7 oz. capacities.
  • Plated aluminum
  • Refillable and rechargeable
  • Working pressure: 80 to 145 psi
  • Measures are the same as aerosol cans (~2³/₄" Ø)
  • Liquid spray lighter than #10 motor oil
All sprayers come standard with Viton® seals, these seals are compatible with most products.
Specifications
  • Description:
    Compete Repair Kit