Bacti-Cinerator IV, Electron Microscopy Sciences
Product Details & Documents
The Baci-Cinerator IV sterilizes by infrared heat (Heater Element contains no asbestos) - organic material is incinerated deep inside the ceramic tunnel. To use, simply insert loop or needle into the tunnel. Complete sterilization takes only 5 to 7 seconds at optimum sterilizing temperature of 1500 °F.
- Prevents infectious spatter and/or cross contamination
- No open flame -no hazardous gas
- No need to wait for the loops or needles to turn red
- Does not require oxygen so it can be used in anaerobic chambers
- UL approved
Specifications
- Description:Bacti-Cinerator IV
- Electrical:220 V
Accessories
Product Information
- Catalog #AvailabilityPrice
- Replacement Switch, Bacti-Cinerator III