Order Entry
United States
Orders LinkContactUsLinkComponent
 

Lanthanum hexaboride ≥99% (La basis)

Glass bottle, 10 g
Catalog # 100205-676
Marketsource
Supplier:   Strem Chemicals Inc
 :  93-5735-10G
Lanthanum hexaboride ≥99% (La basis)
Catalog # 100205-676
Marketsource
CAS Number:  12008-21-8
Lanthanum hexaboride ≥99% (La basis)

Glass bottle, 10 g

CAS Number:  12008-21-8
Price
$76.43
Log in for availability
Each (10g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Lanthanum boride
  • CAS Number:12008-21-8
  • Formula:B₆La
  • MDL Number:MFCD00151350
  • Molecular Weight:203.77
  • Melting Pt:2210 °C
  • Density:2.61
  •  :Density: 2.61 g/cm³ (20 °C)
  • EINECS:234-531-6
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    10 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.