Casein Kinase II
10000 units
Catalog # 101641-528
Small Business Enterprise
Supplier: New England Biolabs (NEB)
Supplier #: P6010S
Casein Kinase II
Catalog # 101641-528
Supplier: New England Biolabs (NEB)
Small Business Enterprise
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Casein Kinase II (CK2) is a constitutively active serine/threonine protein kinase composed of two 44 kDa catalytic α-subunits and two 26 kDa regulatory β-subunits in an α2β2 configuration to form stable heterotetramers
CK2 holoenzyme undergoes autophosphorylation at two serine residues (S2/S3) of its β-subunit. Recently it has been shown that CK2 α-subunits undergo intermolecular tyrosine-autophosphorylation at Y182, which may represent a specific regulatory mechanism. Also, CK2 is able to phosphorylate, under special circumstances, tyrosyl residues in proteins. CK2 is implicated in a variety of cellular functions.
Specifications
Specifications
- Size:10000 units
- Enzyme Synonyms:CKII
- Cat. No.:101641-528
- Enzyme Name:Casein Kinase II