IMO Dangerous Goods Declaration Form, Labelmaster
1-part blank laser form, PP/cellulose, 8.5", 11"
Catalog # 101378-096
Marketsource
Supplier: Label Master
Supplier #: F03L-B
IMO Dangerous Goods Declaration Form, Labelmaster
Catalog # 101378-096
Supplier: Label Master
Marketsource
Jump to:
Product Details & Documents
Adhere to the declaration recommended by the International Maritime Organization (IMO) by using the IMO dangerous goods declaration form.
- Blue and white striped border
- Available in multiple formats
This hazmat form is required by shippers who offer dangerous goods for transport on maritime carriers like cargo and passenger ships. It requires specific information regarding commodity, hazard classification, shipper, receiver, point of origin to destination, and much more. This IMDG Code form may consist of more than one page, provided pages are consecutively numbered. Use Masterform software to fill out the laser forms.
Packaging: Sold in packages of 100 sheets.
Specifications
- Description:1-part blank laser form
- Height:11"
- Material:PP/cellulose
- Width:8.5"



