Order Entry
United States
Orders LinkContactUsLinkComponent
Filter Flasks, Advantec MFS

Filter Flasks, Advantec MFS

Filter Flask, 250 ml, 19/22
Catalog # 101104-936
Marketsource
Supplier:   Advantec MFS
Supplier #:  311225
Filter Flasks, Advantec MFS
Catalog # 101104-936
Supplier:  Advantec MFS
Marketsource
Filter Flasks, Advantec MFS

Filter Flask, 250 ml, 19/22

Price
$285.03
Log in for availability
Pack of 1
Minimum order quantity is 1
Product Details & Documents

These borosilicate filter flasks and silicone rubber stoppers are compatible with Advantec filter holders.

  • Side arm connection 3/8"
  • Flasks with ground glass joint fit holders with female joint (GFA, P/FC 2)

These flasks can be used to collect filtrate or to act as a trap to prevent liquid from damaging pump or vacuum line.

#5 and #8b stoppers adapt 13 mm, 25 mm and KP 47 filter holders to 125 ml (#5) and 1 l (#8b) flasks. #8 stopper adapts 47 mm (except KP 47) and 90 mm filter holders to the 1 l flask.

Specifications
  • [ST] joint no.:
    19/22
  • Capacity:
    250 ml
  • Description:
    Filter Flask