Orders LinkContactUsLinkComponent
Perfect Scale, Electron Microscopy Sciences
Scale only, Perfect
  100493-002
 :  70617-20
Perfect Scale, Electron Microscopy Sciences
  100493-002
Perfect Scale, Electron Microscopy Sciences
Perfect Scale, Electron Microscopy Sciences
Scale only, Perfect
$129.42
Each
The Perfect scale is made from Nickel alloy and 3 mm in length with 1/10 mm divisions. The Scale fits the Perfect Handle (70616-HD).
 : The Perfect Scale comes complete with one scale and a handle.
  • Description:
    Scale only, Perfect