Light Boxes:, Electron Microscopy Sciences
Product Details & Documents
Model A: It is compact and can be used on a desk
For viewing and editing slides, transparencies, negatives, micrographs, etc. They also serve as a base for orientation of embedding specimens when observing tissues using a magnifier. Scientifically designed to provide even distribution and uniformity of light. All metal welded construction with a white baked-enamel finish. The viewing surface is a translucent acrylic that is break resistant. It can be used flat on a table, inclined, standing, or hung on the wall. It comes with a ANSI standard 5000K fluorescent lamp, 10 feet cord, on-off switch.
Model A: It is compact and can be used on a desk. Small enough to carry in a briefcase or store in a drawer. Weight 4.5 lbs. Viewing surface measures: 22 x 29.2 cm (8 3/4 x 11 1/2"). Overall measurements: 24.1 x 31.6 x 5 cm (9 1/2 x 12 1/2 x 2").
Small enough to carry in a briefcase or store in a drawer. Weight 4.5 lbs. Viewing surface measures: 22 x 29.2 cm (8 3/4 x 11 1/2"). Overall measurements: 24.1 x 31.6 x 5 cm (9 1/2 x 12 1/2 x 2").
Specifications
- Description:Light Box, Model A