AMRAY 1000/1200 SEM Mounts, Electron Microscopy Sciences
Product Details & Documents
Head: 25.4 (1") diameter. Pin 3.1 mm (1/8") diameter.
Specifications
- Description:Amray 1000/1200
- Material:Aluminum