Order Entry
United States
Orders LinkContactUsLinkComponent
pH Buffer Solutions, Aqua Solutions

pH Buffer Solutions, Aqua Solutions

pH Buffer Solution, 20 L, Plastic bottle, clear, 20 L, 8
Catalog # 101205-962
Marketsource
Supplier:   Aqua Solutions
Supplier #:  1510-20L
pH Buffer Solutions, Aqua Solutions
Catalog # 101205-962
Supplier:  Aqua Solutions
Marketsource
CAS Number:  
pH Buffer Solutions, Aqua Solutions
pH Buffer Solutions, Aqua Solutions

pH Buffer Solution, 20 L, Plastic bottle, clear, 20 L, 8

CAS Number:  
Price
$205.74
Log in for availability
Each (20l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Specifications

Specifications

  • Pack type:
    Plastic bottle, clear
  • Description:
    pH Buffer Solution
  • Size:
    20 L
  • pH:
    8
  • Volume:
    20 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.