Order Entry
United States
Orders LinkContactUsLinkComponent
Sorensen's Phosphate Buffer, Electron Microscopy Sciences

Sorensen's Phosphate Buffer, Electron Microscopy Sciences

Sorensons Phosphate Buffer, 0.2 M, 1 L, 2 x 1 mm Slot, 7.2
Catalog # 100496-400
Minority or Woman-Owned Business Enterprise
Supplier #:  11600-10
Sorensen's Phosphate Buffer, Electron Microscopy Sciences
Catalog # 100496-400
Minority or Woman-Owned Business Enterprise
CAS Number:  
Sorensen's Phosphate Buffer, Electron Microscopy Sciences
Sorensen's Phosphate Buffer, Electron Microscopy Sciences

Sorensons Phosphate Buffer, 0.2 M, 1 L, 2 x 1 mm Slot, 7.2

CAS Number:  
Price
$70.85
Log in for availability
Each (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Sorensen's Phosphate Buffer is also know as phosphate mixed sodium salts. The osmolarity of a 0.1 M at a pH 7.2 is 226 mosmols; with the addition of 0.18 M of sucrose to the solution, the osmolarity raises to 425 mosmols.

  • pH can be specified when ordering
  • Maximum shelf life after receipt is 3 months

Ordering information: Certificates of analysis are available by lot or product number.

Specifications

Specifications

  • Description:
    Sorensons Phosphate Buffer, 0.2 M
  • Size:
    2 x 1 mm Slot
  • pH:
    7.2
  • Volume:
    1 L
  • Cat. No.:
    100496-400
  • Size:
    1 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.