Aldehyde/Silane Slide, Electron Microscopy Sciences
Aldehyde/Silane Slide With ES
Catalog # 101412-858
Jump to:
Discontinued
Product Details & Documents
Offers a uniform coating of aldehydes for attaching amine-modified DNA.
Specifications
- Description:Aldehyde/Silane Slide With ES