Order Entry
United States
Orders LinkContactUsLinkComponent
 

Alcian Blue Method (pH 2.5), Electron Microscopy Sciences

Acetic Acid 3% Aqueous, 500 ml
Catalog # 101411-238
 :  26323-03
Alcian Blue Method (pH 2.5), Electron Microscopy Sciences
Catalog # 101411-238
CAS Number:  
Alcian Blue Method (pH 2.5), Electron Microscopy Sciences
Alcian Blue Method (pH 2.5), Electron Microscopy Sciences

Acetic Acid 3% Aqueous, 500 ml

CAS Number:  
Price
$35.86
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specifications

Specifications

  • Description:
    Acetic Acid 3% Aqueous
  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.