Order Entry
United States
Orders LinkContactUsLinkComponent
 

Collodion solution 5% USP

Bottle, 100 ml
Catalog # 100503-766
Flammable
Irritant
Health
 :  12600
Collodion solution 5% USP
Catalog # 100503-766
CAS Number:  9004-70-0
Collodion solution 5% USP
Collodion solution 5% USP

Bottle, 100 ml

CAS Number:  9004-70-0
Price
$45.94
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

25% Alcohol, 75% Diethyl Ether in Pyroxylin

Electron Microscopy Sciences are now offering Collodion in a 30 ml bottle to avoid its classification as a hazardous chemical for shipping purposes.

  • CAS Number:9004-70-0
  • MDL Number:MFCD00081525
  • Boiling Pt:35 °C (95 °F)
  • Flash Pt:–52 °C (–62 °F)
  • Storage Temperature:Ambient
  • UN:1993
  • ADR:3,I
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    100 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.