Orders LinkContactUsLinkComponent
SS Expandable Capsule, Electron Microscopy Sciences
Expandable Capsule
  100503-074
SS Expandable Capsule, Electron Microscopy Sciences
  100503-074
 :  
SS Expandable Capsule, Electron Microscopy Sciences
SS Expandable Capsule, Electron Microscopy Sciences
Expandable Capsule
Made from stainless steel and comes with a positive snap cap. They are expandable. When closed its diameter by depth dimensions are 40 x 7 mm and can be expanded to 11 mm deep. Perforated top and bottom.
  • Description:
    Expandable Capsule