Orders LinkContactUsLinkComponent
Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Flint, 3.5"L
Catalog # 100490-960
Supplier #:  70950-52
Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Catalog # 100490-960
Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Flint, 3.5"L
Price
$68.21
Log in for availability
Pack of 250
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Good for handling infectious materials, inoculating culture media and routine sampling. Available in either Borosilicate or Flint glass, with an outside body diameter of 7 mm.
Specifications
  • Description:
    Flint
  • Length:
    3.5"L