Order Entry
United States
Orders LinkContactUsLinkComponent
 

Selenium dioxide ≥99.8%

Glass bottle, 100 g
Catalog # 100200-554
Toxic
Health
Environment
Marketsource
Supplier:   Strem Chemicals Inc
 :  93-3406-100G
Selenium dioxide ≥99.8%
Catalog # 100200-554
Marketsource
CAS Number:  7446-08-4
Selenium dioxide ≥99.8%

Glass bottle, 100 g

CAS Number:  7446-08-4
Price
$164.18
Log in for availability
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:7446-08-4
  • Formula:SeO₂
  • MDL Number:MFCD00003562
  • Molecular Weight:110.96 g/mol
  • Merck Index:13,08508
  • Boiling Pt:340…350 °C (subl.)
  • Melting Pt:315 °C
  • Density:3.95
  • Storage Temperature:Ambient
  • EINECS:231-194-7
  • UN:3283
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.