Avery® Custom Binder Spine Inserts
White, Inkjet Printers, Laser Printers, ½"
Catalog # 500015-072
Supplier: Janitorial Supplies
Supplier #: AVE89101
Jump to:
Product Details & Documents
Create professional-looking binder spines using your inkjet or laser printer.
Micro-perforated cardstock for easy separation of each spine. Firm cardstock makes it easier to insert each spine into a binder.
Specifications
- For Use With:Inkjet Printers, Laser Printers
- Color:White
- Width:½"



