Order Entry
United States
Orders LinkContactUsLinkComponent
Pacon® Array® Card Stock

Pacon® Array® Card Stock

Card stock, Black, 8.5×11", 65 lbs.
Catalog # 500004-262
Supplier:   Janitorial Supplies
Supplier #:  PAC101187
Pacon® Array® Card Stock
Catalog # 500004-262
Pacon® Array® Card Stock
Pacon® Array® Card Stock

Card stock, Black, 8.5×11", 65 lbs.

Price
$16.63
Log in for availability
Pack of 100
Minimum order quantity is 1
Product Details & Documents

For report covers, flyers, postcards and art projects.

  • Acid-free for archival quality

Used in printers, copiers or typewriters.

Specifications
  • Color:
    Black
  • Description:
    Card stock
  • Dimensions:
    8.5×11"
  • Weight:
    65 lbs.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.