Orders LinkContactUsLinkComponent
SuperFrost™ Clipped Corner Slide, Electron Microscopy Sciences
SuperFrost™ Clipped Corner Slide, Electron Microscopy Sciences
Superfrost clipped corner, 1 g
Catalog # 101413-468
Supplier #:  71882-01
SuperFrost™ Clipped Corner Slide, Electron Microscopy Sciences
Catalog # 101413-468
SuperFrost™ Clipped Corner Slide, Electron Microscopy Sciences
SuperFrost™ Clipped Corner Slide, Electron Microscopy Sciences
Superfrost clipped corner, 1 g
Price
$104.67
Log in for availability
Pack of 144
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Designed specially for the Sysmex SP-100 slide prep unit and recommended for the Diagnostics.
The 45 degree clipped corner slide offers exceptional cleanliness and resistance to sticking. The tight tolerances for the length and width help to reduce accumulation of glass chips within the instrument. The pre-cleaned slide is 25×75 mm and approximately 1 mm thick. Ideal for technicians, who routinely prepare large number of slides.
Specifications
  • Capacity:
    1 g
  • Description:
    Superfrost clipped corner