Order Entry
United States
Orders LinkContactUsLinkComponent
BssHII

BssHII

BssHII, 5000 units/ml, 2500 units
Catalog # 101229-012
Small Business Enterprise
Supplier #:  R0199L
BssHII
Catalog # 101229-012
Small Business Enterprise
CAS Number:  
BssHII
BssHII

BssHII, 5000 units/ml, 2500 units

CAS Number:  
Price
$343.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

An E.coli strain that carries the BssHII gene from Bacillus stearothermophilus H3 (N. Welker).

Specifications

Specifications

  • Concentration:
    5000 units/ml
  • Description:
    BssHII
  • Size:
    2500 units
  • Cat. No.:
    101229-012
  • Enzyme Name:
    BssHII

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.