Embedding Rings, Electron Microscopy Sciences
Product Details & Documents
Molded from the highest quality plastic, our Embedding Rings fit most base molds. For added convenience, there are four colors from which to choose Blue, Pink, Green, Yellow. The large writing surface further eases identification and handling. The base measures 1 5/8 x 1 1/8" and the ring measures 25 x 25 mm (1 x 1").
Specifications
- Color:Blue
- Description:Embedding ring