Order Entry
United States
Orders LinkContactUsLinkComponent
 

HON® Task Light for Stack-on Storage Unit

Fluorescent light T5 for HON® 48" Wide Stack-On Storage Unit, 34⅝", 1⅛", 4⅞"
Catalog # 500016-634
Supplier:   Janitorial Supplies
 :  HONH870942
HON® Task Light for Stack-on Storage Unit
Catalog # 500016-634
HON® Task Light for Stack-on Storage Unit
HON® Task Light for Stack-on Storage Unit

Fluorescent light T5 for HON® 48" Wide Stack-On Storage Unit, 34⅝", 1⅛", 4⅞"

Price
$183.32
Log in for availability
Each
 
Product Details & Documents

Provides additional light to your work area.

  • Slim profile design mounts recessed under Stack-on Storage Unit
  • Mounts with ease with steel spring clips
  • Electronic ballast for increased energy efficiency
  • 3 prong plug and black base
  • SLT-30534 replacement bulb for 48"; SLT-01715 replacement bulb for 72" lamp

9' cord

 : UL Approved.

 : Bulb included.

Specifications
  • Description:
    Fluorescent light T5 for HON® 48" Wide Stack-On Storage Unit
  • Height:
    1⅛"
  • Depth:
    4⅞"
  • Width:
    34⅝"