Order Entry
United States
Orders LinkContactUsLinkComponent
 

Universal® Quick Set-up String-and-Button Boxes

White, Corrugated Fiberboard, 510 lbs., Letter, 12×24×10" (Inner)
Catalog # 500003-171
   Sustainable Materials
Supplier:   Janitorial Supplies
 :  UNV75121
Universal® Quick Set-up String-and-Button Boxes
Catalog # 500003-171
   Sustainable Materials
Universal® Quick Set-up String-and-Button Boxes
Universal® Quick Set-up String-and-Button Boxes

White, Corrugated Fiberboard, 510 lbs., Letter, 12×24×10" (Inner)

Price
$180.65
Log in for availability
Carton of 12
 
Product Details & Documents

Save assembly time with this light-duty box.

Sturdy corrugated construction for strength and value. String-and-button tie closure securely stores box contents.

Specifications
  • Color:
    White
  • Dimensions:
    12×24×10" (Inner)
  • Material:
    Corrugated Fiberboard
  • Size:
    Letter
  • Stacking strength:
    510 lbs.
  • Cat. No.:
    500003-171
  • Environmentally Preferable:
    Sustainable Materials