Orders LinkContactUsLinkComponent
Aldehyde/Silane Slide, Electron Microscopy Sciences
Aldehyde/Silane Slide, Electron Microscopy Sciences
Aldehyde/Silane Slide With ES & Bar Code
Catalog # 101412-856
Supplier #:  63476-EB
Aldehyde/Silane Slide, Electron Microscopy Sciences
Catalog # 101412-856
Aldehyde/Silane Slide, Electron Microscopy Sciences
Aldehyde/Silane Slide, Electron Microscopy Sciences
Aldehyde/Silane Slide With ES & Bar Code
Price
$729.60
Log in for availability
Pack of 20
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Offers a uniform coating of aldehydes for attaching amine-modified DNA.
Specifications
  • Description:
    Aldehyde/Silane Slide With ES & Bar Code