Orders LinkContactUsLinkComponent
Polyethylene Molds, Embedding Stubs, Electron Microscopy Sciences
Polyethylene Molds, Embedding Stubs, Electron Microscopy Sciences
Stubs Thick Embedding 6 mm
Catalog # 100502-820
Supplier #:  70175-50
Polyethylene Molds, Embedding Stubs, Electron Microscopy Sciences
Catalog # 100502-820
Polyethylene Molds, Embedding Stubs, Electron Microscopy Sciences
Polyethylene Molds, Embedding Stubs, Electron Microscopy Sciences
Stubs Thick Embedding 6 mm
Price
$276.91
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Block holders molded from hard plastic are easy to store as it attched to Aluminum chucks (ABH)
The large center hole in the holder prevents bubble formation and its frosted surface is easy for marking.
Specifications
  • Description:
    Stubs Thick Embedding 6 mm