Orders LinkContactUsLinkComponent
DrainRack™ Slide Rack, Electron Microscopy Sciences
DrainRack™ Slide Rack, Electron Microscopy Sciences
Green
Catalog # 100490-988
Supplier #:  71446-G
DrainRack™ Slide Rack, Electron Microscopy Sciences
Catalog # 100490-988
DrainRack™ Slide Rack, Electron Microscopy Sciences
DrainRack™ Slide Rack, Electron Microscopy Sciences
Green
Price
$73.53
Log in for availability
Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from high impact polystyrene. This rugged tray used as a drain rack can hold up to 200 microscope slides in 100 individual numbered slots.
All sides are stored upright for easy insertion and removal. A unique feature with the DrainRack™ is the ability to read bar codes without having to remove the slides from the rack. For space saving purposes, you can double the amount of slides simply by storing 2 slides per slot, giving you a capacity of 200 slides per tray instead of 100. The racks are not autoclavable. They measure: 75 x 231 x 25 mm (3 x 9 x 1").
Specifications
  • Color:
    Green