Orders LinkContactUsLinkComponent
Petri Dishes, Disposable, Electron Microscopy Sciences
Petri Dishes, Disposable, Electron Microscopy Sciences
Quad-Petri Dish
Catalog # 100488-410
Supplier #:  70686
Petri Dishes, Disposable, Electron Microscopy Sciences
Catalog # 100488-410
Petri Dishes, Disposable, Electron Microscopy Sciences
Petri Dishes, Disposable, Electron Microscopy Sciences
Quad-Petri Dish
Price
$114.35
Log in for availability
Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Our petri dishes are available in a variety of sizes and shapes for use in routine procedures and automatic equipmentF. Manufactured from Polystyrene, with excellent optical clarity. The dishes are sterile and each lot is tested to assure uniform quality and sterility. Sterlstar Petri Dishes are designed for the Microbiologist who prefers a heavier dish. The Sterlstar dish offers easy handling, a secure grip, and additional strength.
Specifications
  • Description:
    Quad-Petri Dish