Order Entry
United States
Orders LinkContactUsLinkComponent
Matrix Printer Labels, Dots, Bulk Pack, Universal

Matrix Printer Labels, Dots, Bulk Pack, Universal

Matrix printer labels, dots, bulk pack, Paper, White, Dot matrix printers, 1.44×4"
Catalog # 500016-220
Matrix Printer Labels, Dots, Bulk Pack, Universal
Catalog # 500016-220
Supplier:  
Matrix Printer Labels, Dots, Bulk Pack, Universal
Matrix Printer Labels, Dots, Bulk Pack, Universal

Matrix printer labels, dots, bulk pack, Paper, White, Dot matrix printers, 1.44×4"

Discontinued
Product Details & Documents

Print labels for mass mailings from your pin-fed printer.

  • Perforated at the page fold after every 12" across one label
  • Rectangular shape

Self-adhesive labels apply quickly.

Specifications
  • For Use With:
    Dot matrix printers
  • Color:
    White
  • Description:
    Matrix printer labels, dots, bulk pack
  • Dimensions:
    1.44×4"
  • Material:
    Paper
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.