Order Entry
United States
Orders LinkContactUsLinkComponent
Matrix Printer Labels, Dots, Bulk Pack, Universal

Matrix Printer Labels, Dots, Bulk Pack, Universal

Matrix printer labels, dots, bulk pack, Paper, White, Dot matrix printers, 0.94×3.5"
Catalog # 500016-208
Supplier:   Janitorial Supplies
Supplier #:  UNV70104
Matrix Printer Labels, Dots, Bulk Pack, Universal
Catalog # 500016-208
Matrix Printer Labels, Dots, Bulk Pack, Universal
Matrix Printer Labels, Dots, Bulk Pack, Universal

Matrix printer labels, dots, bulk pack, Paper, White, Dot matrix printers, 0.94×3.5"

Price
$27.71
Log in for availability
Box of 5,000
Minimum order quantity is 1
Product Details & Documents

Print labels for mass mailings from your pin-fed printer.

  • Perforated at the page fold after every 12" across one label
  • Rectangular shape

Self-adhesive labels apply quickly.

Specifications
  • For Use With:
    Dot matrix printers
  • Color:
    White
  • Description:
    Matrix printer labels, dots, bulk pack
  • Dimensions:
    0.94×3.5"
  • Material:
    Paper