Avery® Custom Binder Spine Inserts
White, Inkjet Printers, Laser Printers, 2"
Catalog # 500001-566
Supplier: Janitorial Supplies
Supplier #: AVE89107
Jump to:
Product Details & Documents
Create professional-looking binder spines using your inkjet or laser printer.
Micro-perforated cardstock for easy separation of each spine. Firm cardstock makes it easier to insert each spine into a binder.
Specifications
- For Use With:Inkjet Printers, Laser Printers
- Color:White
- Width:2"
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



