Microtube Rack - Microwaveable, Electron Microscopy Sciences
Polypropylene floating racks keep microtubes submerged in a water bath – perfect for controlling sample temperature. Detachable legs give the users more options: attach legs so rack stands upright on benchtop, or detach legs so sample tubes can be quickly popped out by pushing down onto any surface. Ideally to hold microcentrifuge tube in water bath during polymerization, such as LR White®. Not autoclavable. Measures: 60 mm high including legs.
- Description:Rack Micro Tube Sq white