Orders LinkContactUsLinkComponent
Mcpherson Tying Forceps, Electron Microscopy Sciences
Mcpherson Tying Forceps, Electron Microscopy Sciences
Mcpherson Tying Forceps, Straight
Catalog # 100494-920
Supplier #:  78615-4S
Mcpherson Tying Forceps, Electron Microscopy Sciences
Catalog # 100494-920
Mcpherson Tying Forceps, Electron Microscopy Sciences
Mcpherson Tying Forceps, Electron Microscopy Sciences
Mcpherson Tying Forceps, Straight
Price
$317.97
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Available with straight or angled tips with 5 mm tying platform (parallel gripping platform). Measures 4" long.
Specifications
  • Description:
    Mcpherson Tying Forceps, Straight